Advantech L3-S18 Standard Sieve, 3" dia, Stainless Steel Mesh, 1.00 mm Sieve Opening, No. 18
Representative image only.
Advantech Mfr # L3-S18 – Item # WZ-59948-30
Steel mesh meets ASTM and ISO specifications
  • 1 year warranty
PRODUCT OPTIONS
£139.17GBP / Each
£167.00GBP (incl VAT)
£126.63 - £185.17GBP / Each
£151.96 - £222.20GBP (incl VAT)
for an additional
£126.63GBP / Each
£151.96GBP (incl VAT)
for an additional
£185.17GBP / Each
£222.20GBP (incl VAT)
Time to recalibrate? Click on the calibration service to view details, then order.
SPECS & DESCRIPTION
Specifications & Description
  • MaterialAcrylic frame, stainless steel wire
  • U.S. Standard Screen Size (No.)18
  • Tyler Standard Screen Size (mesh)16
  • Sieve Opening (mm)1
  • Inside Diameter (InchShort)3
  • Height (InchShort)0.625
  • DescriptionStandard Sieve, Stainless Steel Mesh, 1.00 mm sieve opening, No. 18
  • Warranty1 Year
1 Year Warranty
Key Features
  • 1 year warranty
More About this Item
Standard sieves with stainless steel mesh meet ASTM and ISO specifications and are supplied with a certificate of compliance. Sieves measure 3" ID and 5/8" H.