This item is now discontinued. Please browse similar items
Advantec PG60110MM Air Sampling Filters, Composite, 110-mm dia disc; pack of 50.
Representative image only.
Advantec Mfr # PG60110MM – Item # WZ-06649-62
Composite filters ideal for sampling airborne particulates
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • MaterialPTFE
  • Diameter (mm)110
  • Diameter (cm)11
  • DescriptionAir Sampling Filters, Composite, 110-mm dia disc; pack of 50.
  • Warranty1 year
1 Year Warranty
Key Features
  • 1 year warranty
More About this Item
Composite Filters are specially developed for the measurement of dusts and particulate matter in the air. These PTFE coated filters are naturally hydrophobic and unaffected by humidity. Thermo-stable to 500°F (260°C).