This item is now discontinued. We recommend the following as suggested replacements: WZ-60061-78
Advantec M-08537MM Cellulose pad for air-monitoring cassette, 37-mm diameter; pack of 100
Representative image only.
Advantec Mfr # M-08537MM – Item # WZ-02935-36
Assemble you own monitor for specific applications
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • Filter Diameter (mm)37
  • DescriptionCellulose pad for air-monitoring cassette, 37-mm diameter; pack of 100
  • Warranty1 year
1 Year Warranty
Key Features
  • 1 year warranty
More About this Item
Empty cassettes and cellulose pads are available as an option for you to assemble a filter/monitoring unit with an MCE membrane filter for asbestos monitoring or other 37-mm filter.