Advantec GC90125MM Grade GC90 Glass Fiber Filters; 125mm dia; 100/box
Representative image only.
Advantec Mfr # GC90125MM – Item # WZ-06657-86
For analytical or gravimetric studies
  • 1 year warranty
PRODUCT OPTIONS
£257.97GBP / Pkg of 100
£309.56GBP (incl VAT)
SPECS & DESCRIPTION
Specifications & Description
  • MaterialGlass Microfiber
  • Diameter (mm)125
  • Diameter (cm)12.5
  • DescriptionGrade GC90 Glass Fiber Filters; 125mm dia; 100/box
  • Warranty1 year
1 Year Warranty
Key Features
  • 1 year warranty
More About this Item
High wet strength.