This item is now discontinued. Please browse similar items
Advantec GB100R55MM Glass fiber filter, GB100R, 55-mm dia; box of 100
Representative image only.
Advantec Mfr # GB100R55MM – Item # WZ-06657-73
For analytical or gravimetric studies
  • Borosilicate glass filters create a biologically inert filter matrix that is resistant to both chemicals and pH extremes
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • MaterialGlass Microfiber
  • Diameter (mm)55
  • Diameter (cm)5.5
  • DescriptionGlass fiber filter, GB100R, 55-mm dia; box of 100
  • Warranty1 year
1 Year Warranty
Key Features
  • Borosilicate glass filters create a biologically inert filter matrix that is resistant to both chemicals and pH extremes
  • 1 year warranty
More About this Item
Use as prefilters to extend membrane life, for water/air pollution analysis, or liquid clarification. Filters without binder withstand up to 932°F (500°C); filters with binder withstand up to 250°F (120°C). Grades GC90, GS25, and DP70 with binder offer higher wet strength and lower fiber release. Quartz fiber filters have very low trace metal content and can withstand up to 1832°F (1000°C). Choose quartz fiber filters to sample acidic gases at high temperatures or for air pollution analysis.