This item is now discontinued. Please browse similar items
Advantec GB100R293MM Grade GB100R Glass Fiber Filters; 293mm dia; 50/box
Representative image only.
Advantec Mfr # GB100R293MM – Item # WZ-06657-78
For analytical or gravimetric studies
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • MaterialGlass Microfiber
  • Diameter (mm)293
  • Diameter (cm)29.3
  • DescriptionGrade GB100R Glass Fiber Filters; 293mm dia; 50/box
  • Warranty1 year
1 Year Warranty
Key Features
  • 1 year warranty
More About this Item
Low trace metal content; sampling high and low volume aerosols for airborne dust and metal contaminants.