This item is now discontinued. Please browse similar items
Advantec GA200125MM Grade GA200 Glass Fiber Filters; 125mm dia; 50/box
Representative image only.
Advantec Mfr # GA200125MM – Item # WZ-06657-66
For analytical or gravimetric studies
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • MaterialGlass Microfiber
  • Diameter (mm)125
  • Diameter (cm)12.5
  • DescriptionGrade GA200 Glass Fiber Filters; 125mm dia; 50/box
  • Warranty1 year
1 Year Warranty
Key Features
  • 1 year warranty
More About this Item
Thick Filter; filtering viscous fluids such as liquid sugars and gels.