This item is now discontinued. Please browse similar items
Advantec GA100293MM Grade GA100 Glass Fiber Filters; 293mm dia; 50/box
Representative image only.
Advantec Mfr # GA100293MM – Item # WZ-06657-58
For analytical or gravimetric studies
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • MaterialGlass Microfiber
  • Diameter (mm)293
  • Diameter (cm)29.3
  • DescriptionGrade GA100 Glass Fiber Filters; 293mm dia; 50/box
  • Warranty1 year
1 Year Warranty
Key Features
  • 1 year warranty
More About this Item
General purpose; filtering cells and precipitated proteins; air pollution monitoring.