This item is now discontinued. Please browse similar items
Advantec DP70293MM Grade DP70 Glass Fiber Filters; 293mm dia; 50/box
Representative image only.
Advantec Mfr # DP70293MM – Item # WZ-06658-08
For analytical or gravimetric studies
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • MaterialGlass Microfiber
  • Diameter (mm)293
  • Diameter (cm)29.3
  • DescriptionGrade DP70 Glass Fiber Filters; 293mm dia; 50/box
  • Warranty1 year
1 Year Warranty
Key Features
  • 1 year warranty
More About this Item
High wet strength and very high loading capacity; dust and fine particulate sampling.