Advantec DP70125MM Grade DP70 Glass Fiber Filters; 125mm dia; 50/box
Representative image only.
Advantec Mfr # DP70125MM – Item # WZ-06658-06
For analytical or gravimetric studies
PRODUCT OPTIONS
£121.80GBP / Pkg of 50
£146.16GBP (incl VAT)
SPECS & DESCRIPTION
Specifications & Description
  • MaterialGlass Microfiber
  • Diameter (mm)125
  • Diameter (cm)12.5
  • DescriptionGrade DP70 Glass Fiber Filters; 125mm dia; 50/box
More About this Item
High wet strength and very high loading capacity; dust and fine particulate sampling.