This item is now discontinued. Please browse similar items
Advantec CCP-3-C1H Disposable Capsule Filters, Polypropylene Depth-Media; 5-10 µm
Representative image only.
Advantec Mfr # CCP-3-C1H – Item # WZ-29702-15
Polypropylene (PP) housing available with PTFE or PES membrane, or PP depth media
  • All-polypropylene housing is thermally fused and offers broad chemical compatibility; diameter is a standard 78 mm
  • Capsules are sterilizable by autoclave or ethylene oxide (EtO);
  • Max operating pressure is 57 psi (3.9 bar); max operating temperature is 140°F (60°C)
  • Capsules are vented at both ends for bleeding trapped air and for draining
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • Membrane MaterialPolypropylene
  • Pore Size (micron)1.0
  • Filter Size78
  • Housing MaterialPolypropylene
  • Connections3/8\" hose barb
  • Effective Filtration Area (cm²)600
  • Surface Area (m²)0.06
  • Particle Retention (µm)10
  • Particle Retention (%)98
  • Max Operating Temperature (° C)60
  • Max Operating Temperature (° F)140
  • Max Pressure (PSI)57
  • Max Pressure3.9
  • DescriptionDisposable Capsule Filters, Polypropylene Depth-Media; 5-10 µm
  • Warranty1 year
1 Year Warranty
Key Features
  • All-polypropylene housing is thermally fused and offers broad chemical compatibility; diameter is a standard 78 mm
  • Capsules are sterilizable by autoclave or ethylene oxide (EtO);
  • Max operating pressure is 57 psi (3.9 bar); max operating temperature is 140°F (60°C)
  • Capsules are vented at both ends for bleeding trapped air and for draining
  • 1 year warranty
More About this Item
Polypropylene Depth-Media Capsules offer high dirt holding capacity with superior retention. PP fibers are thermally welded to prevent slough-off and to minimize changes in pore size. Use these filters for high purity water and prefiltration applications.