This item is now discontinued. Please browse similar items
Advantec CCF-050-D1H Disposable Capsule Filters, PTFE Hydrophobic; 0.5 µm; 1150 cm2
Representative image only.
Advantec Mfr # CCF-050-D1H – Item # WZ-29702-25
Polypropylene (PP) housing available with PTFE or PES membrane, or PP depth media
  • All-polypropylene housing is thermally fused and offers broad chemical compatibility; diameter is a standard 78 mm
  • Capsules are sterilizable by autoclave or ethylene oxide (EtO);
  • Max operating pressure is 57 psi (3.9 bar); max operating temperature is 140°F (60°C)
  • Capsules are vented at both ends for bleeding trapped air and for draining
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • Membrane MaterialPTFE Hydrophobic
  • Pore Size (micron)0.5
  • Filter Size78
  • Housing MaterialPolypropylene
  • Connections3/8\" hose barb
  • Effective Filtration Area (cm²)1150
  • Surface Area (m²)0.115
  • Max Operating Temperature (° C)60
  • Max Operating Temperature (° F)140
  • Max Pressure (PSI)57
  • Max Pressure3.9
  • DescriptionDisposable Capsule Filters, PTFE Hydrophobic; 0.5 µm; 1150 cm2
  • Warranty1 year
1 Year Warranty
Key Features
  • All-polypropylene housing is thermally fused and offers broad chemical compatibility; diameter is a standard 78 mm
  • Capsules are sterilizable by autoclave or ethylene oxide (EtO);
  • Max operating pressure is 57 psi (3.9 bar); max operating temperature is 140°F (60°C)
  • Capsules are vented at both ends for bleeding trapped air and for draining
  • 1 year warranty
More About this Item
PTFE Membrane Capsules are ideal for high-purity water and media and also uniquely suitable for corrosive fluids and gases. The hydrophobic nature of the membrane and chemical resistance make PTFE capsules ideal for solvent filtration.