This item is now discontinued. Please browse similar items
Advantec CAPS PTFE 0.50 102MM/570CM2
Representative image only.
Advantec Mfr # CCF-050-C1B – Item # WZ-29705-17
Polypropylene (PP) housing available with PTFE or PES membrane, or PP depth media
  • All-polypropylene housing is thermally fused and offers broad chemical compatibility; diameter is a standard 78 mm
  • Capsules are sterilizable by autoclave or ethylene oxide (EtO);
  • Max operating pressure is 57 psi (3.9 bar); max operating temperature is 140°F (60°C)
  • Capsules are vented at both ends for bleeding trapped air and for draining
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
1 Year Warranty
Key Features
  • All-polypropylene housing is thermally fused and offers broad chemical compatibility; diameter is a standard 78 mm
  • Capsules are sterilizable by autoclave or ethylene oxide (EtO);
  • Max operating pressure is 57 psi (3.9 bar); max operating temperature is 140°F (60°C)
  • Capsules are vented at both ends for bleeding trapped air and for draining