Advantec 315110 Glass Microanalysis Filter Holder, 47 mm, SS Support; 1000 mL
Representative image only.
Advantec Mfr # 315110 – Item # WZ-06644-95
Ideal for small to medium volumes of liquid
  • Standard sizes for microbiology and particulate analysis
  • Select glass support for low pH solutions or for slower flow rates; use stainless for proteinaceous or viscous solutions; use PTFE support for acidic solutions at higher flow rates
  • 1 year warranty
PRODUCT OPTIONS
£623.87GBP / Each
£748.64GBP (incl VAT)
SPECS & DESCRIPTION
Specifications & Description
  • Filter Diameter (mm)47
  • Body MaterialStainless Steel
  • Funnel Size (mL)1000
  • Filtration Area (cm²)9.6
  • SupportStainless Steel
  • Stopper SizeNo. 8
  • DescriptionGlass Microanalysis Filter Holder, 47 mm, SS Support; 1000 mL
  • Warranty1 year
1 Year Warranty
Key Features
  • Standard sizes for microbiology and particulate analysis
  • Select glass support for low pH solutions or for slower flow rates; use stainless for proteinaceous or viscous solutions; use PTFE support for acidic solutions at higher flow rates
  • 1 year warranty
More About this Item
Microanalysis filter holders feature borosilicate glass funnel and base, silicone rubber stopper, and anodized aluminum clamp. The 90-mm holders are ideal for large-volume viscous samples or those with a heavy particle or bioburden that would clog a 47-mm filter.