Advantec 07030051 Test Strips, Chrome Check A (Cr6+), 0 to 50 ppm; 25 Strips/Pk
Representative image only.
Advantec Mfr # 07030051 – Item # WZ-59201-18
Color-change test papers give rapid indication of pH level ion presence
  • Obtain quick ion concentration measurements in the field or laboratory
  • Dip and compare color change — no calibration, electricity, or warm up needed
PRODUCT OPTIONS
£12.75GBP / Pkg of 25
£15.30GBP (incl VAT)
SPECS & DESCRIPTION
Specifications & Description
  • TestChrome
  • Measurement Points0 to 50 ppm
  • Reagent PlatformStrips
  • Tests Per Kit25
  • DescriptionTest Strips, Chrome Check A (Cr6+), 0 to 50 ppm; 25 Strips/Pk
Key Features
  • Obtain quick ion concentration measurements in the field or laboratory
  • Dip and compare color change — no calibration, electricity, or warm up needed
More About this Item
These ion test strips make water testing fast and easy. Conveniently packed in screw top bottles, the test papers are safe even in rugged field applications. Obtaining simultanious pH and ion concentration results couldn't be easier. Simply dip the test strip in your water sample, wait about 15 seconds for the water to penetrate the strip, and compare the color changes against the chart on the bottle's label.