This item is now discontinued. Please browse similar items

Admiral 2511B-75 Air Aspiration System, 0.39 cfm, 21.3"Hg, 120 VAC

Representative image only.
Admiral Item # WZ-79202-20
Compact, lightweight portable aspiration system
  • Dial vacuum gauge and regulator to monitor and adjust vacuum level
  • Liquid trap to avoid ingestion of fluids into the pump
  • Automatic flow shut-off when full
  • 1 year warranty

SPECS & DESCRIPTION

Specifications & Description

  • Wetted PartsGlass-filled Polyester, Ryton®, Viton®, PTFE, Delrin®
  • Free-Air Capacity (CFM)0.39
  • Free-Air Capacity (LPM)11
  • Free-Air Capacity (LPH)660
  • Max Flow Rate (CFM)0.39
  • Max Flow Rate (Liters/min)11
  • Max Vacuum (in Hg)21.3
  • Max Vacuum (mbar)292.2
  • Max Vacuum (mm Hg)219
  • Max Vacuum (torr)219
  • Port Size3/16\" hose barb
  • Max Temperature (° F)104
  • Max Temperature (° C)40
  • Duty CycleContinuous
  • Power (VAC)120
  • Power (amps)1.6
  • Motor TypeShaded pole
  • Height (in)10
  • Length (in)11
  • Height (cm)25.4
  • Length (cm)27.94
  • Height (mm)254
  • Length (mm)279.4
  • Motor hp1-1/4
  • DescriptionAir Aspiration System, 0.39 cfm, 21.3\"Hg, 120 VAC
  • Warranty1 Year
1 Year Warranty

Key Features

  • Dial vacuum gauge and regulator to monitor and adjust vacuum level
  • Liquid trap to avoid ingestion of fluids into the pump
  • Automatic flow shut-off when full
  • 1 year warranty

More About this Item

Pump features an autoclavable collection receiver made of polypropylene, a hydrophobic inline filter to prevent introduction of the aspirate into the pump and automatic shutoff prevent overflow of the aspirate to protect you and the pump from exposure to the media. The pump also features a vacuum regulator to control the flow of liquid. Recommended for aspirating aqueous solutions including buffers, but not for acidic, basic, or organic vapors and gases. 70202-30 and 79202-35 suitable for bleach solutions.

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At+44 (0)1480 272279
Live chat