Ace Glass 7231-65 Separatory Glass Funnel, 45/50 joint, 2000 mL
Representative image only.
Ace Glass Mfr # 7231-65 – Item # WZ-34731-10
Squibb type with plug and lock-tight stopper
  • Leak-tight locking stopper
  • Stopper secured in top joint via cap and O-ring
PRODUCT OPTIONS
£544.53GBP / Each
£653.44GBP (incl VAT)
SPECS & DESCRIPTION
Specifications & Description
  • MaterialGlass
  • Capacity (mL)2000
  • Capacity (oz)64
  • Stopcock Plug Size6
  • Joint Size Number45/50
  • DescriptionSeparatory Glass Funnel, 45/50 joint, 2000 mL
Key Features
  • Leak-tight locking stopper
  • Stopper secured in top joint via cap and O-ring
More About this Item
Funnels feature a leak-tight locking stopper that will not fall out while shaking or transporting. Outer ground joint at top is externally threaded so the stopper can be secured with a cap and O-ring.