Ace Glass 7231-63 Separatory Glass Funnel, 24/40 joint, 1000 mL
Representative image only.
Ace Glass Mfr # 7231-63 – Item # WZ-34731-08
Squibb type with plug and lock-tight stopper
  • Leak-tight locking stopper
  • Stopper secured in top joint via cap and O-ring
PRODUCT OPTIONS
£438.23GBP / Each
£525.88GBP (incl VAT)
SPECS & DESCRIPTION
Specifications & Description
  • MaterialGlass
  • Capacity (mL)1000
  • Capacity (oz)32
  • Stopcock Plug Size4
  • Joint Size Number24/40
  • DescriptionSeparatory Glass Funnel, 24/40 joint, 1000 mL
Key Features
  • Leak-tight locking stopper
  • Stopper secured in top joint via cap and O-ring
More About this Item
Funnels feature a leak-tight locking stopper that will not fall out while shaking or transporting. Outer ground joint at top is externally threaded so the stopper can be secured with a cap and O-ring.