This item is now discontinued. Please browse similar items
Accumet Excel XL150 pH Benchtop Meter Only
Representative image only.
Accumet Mfr # 13636XL150A – Item # WZ-59331-58
Full-color touch screen provides unmatched ease of use in operation and setup
  • Measure and display up to four inputs simultaneously
  • Real-time graphs provide outstanding detail
  • USB, bidirectional RS-232, Ethernet and LAN connectivity
  • Connect up to two stirring probes to provide mixing without stir plates
  • 3 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • DescriptionpH Benchtop Meter Only
  • Warranty3 year
3 Year Warranty
Key Features
  • Measure and display up to four inputs simultaneously
  • Real-time graphs provide outstanding detail
  • USB, bidirectional RS-232, Ethernet and LAN connectivity
  • Connect up to two stirring probes to provide mixing without stir plates
  • 3 year warranty
More About this Item
The XL-series of meters offer you the versatility of having a research-grade single- or multi-parameter pH, mV, ion, conductivity, and dissolved oxygen meter. User prompts guide you through operation and extensive setup screens allow easily customizable settings. Meters offers 2000 data point memory storage with USB and bidirectional RS-232. Upgradable application software, access to Internet, and connection to LAN network printer provide support. User passwords protect your data from accidental deletion, and let you retain unique calibration and instrument settings.
Order electrode separately. pH mode accepts up to five-point pH calibration with custom or preset buffer groups. Offers calibration and high/low alarms. pH accepts electrodes with a BNC connector.