This item is now discontinued. Please browse similar items
Accumet CP620133 accuCap, glass body, spear tip, gel-filled
Representative image only.
Accumet Mfr # CP620133 – Item # WZ-55500-53
Most accurate pH electrode, as tested by the PTB (the German Federal Physical Technical Institute in Braunschweig)*
  • Precise, reliable, and fast readings
  • About 200 times larger junction than standard electrodes makes it impossible to clog
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • ApplicationsSpear-tip
  • Electrode TypeDouble junction
  • Electrode Body MaterialGlass
  • Reference JunctionSingle pore, capillary
  • Min Temperature (° C)0
  • Max Temperature (° C)80
  • ConnectorBNC
  • DescriptionaccuCap, glass body, spear tip, gel-filled
  • Warranty1 year
1 Year Warranty
Key Features
  • Precise, reliable, and fast readings
  • About 200 times larger junction than standard electrodes makes it impossible to clog
  • 1 year warranty
More About this Item
The accuCap electrodes use a single pore capillary reference junction, about 200 times larger than typical ceramic junctions. The unique junction design provides increased electrolyte flow, resulting in greater contact between sample and reference, ensuring fast, stable readings. The double junction design offers enhanced performance. A diffusion barrier separates the AgCl reservoir from the reference electrolyte, allowing the use of silver-free electrolytes and preventing AgCl loss due to temperature variations. The end result is an extremely stable reference potential, providing long electrode life.
*See "Traceability of pH measurement" by Petra Spitzer; ISBN 3-89429-877-4 or SSN 0947-7063
accuCap Spear Tip Electrode
Rugged spear tip is ideal for soft foods. Using the accuCap Polymer gel electrolyte provides the same reliable performance needed in difficult samples.