This item is now discontinued. Please browse similar items
Accuform ZFD816VP NFPA Notice Hazard Rating, Sign Kit, 14" X 10", plastic
Representative image only.
Accuform Mfr # ZFD816VP – Item # WZ-41010-16
Signs and kit to help you with regulatory facility compliance
SPECS & DESCRIPTION
Specifications & Description
  • HeaderNOTICE
  • LanguagesEnglish
  • MaterialPlastic
  • Width (in)10
  • Height (in)14
  • DescriptionNFPA Notice Hazard Rating, Sign Kit, 14\" X 10\", plastic
More About this Item
These signs display the four-color National Fire Protection Association (NFPA) diamond format. Signs feature blank hazard rating panels — write in the information for your specific use.
Kit contains one sign, NFPA wallet guide, and adhesive numbers and hazard symbols which are printed black on clear adhesive polyester to fit the hazard panels.