This item is now discontinued. Please browse similar items
Accuform ZFD100XT NFPA Placard, Sign Only, 10" X 10", plastic
Representative image only.
Accuform Mfr # ZFD100XT – Item # WZ-41010-26
For on-the-spot classification of potentially hazardous chemicals
SPECS & DESCRIPTION
Specifications & Description
  • HeaderNFPA PLACARD
  • LanguagesEnglish
  • MaterialPlastic
  • Width (in)10
  • Height (in)10
  • DescriptionNFPA Placard, Sign Only, 10\" X 10\", plastic
More About this Item
These National Fire Protection Association (NFPA) Placards offer easy of installation and convenience. Blank placards are printed with NFPA color coding; simply write in the appropriate number rating in each quadrant.