This item is now discontinued. Please browse similar items
Accuform TSP101PTP Lockout/Tagout Tag, Danger-Do Not Operate, Bilingual, 5.8x3.4", Plastic; 25/Pk
Representative image only.
Accuform Mfr # TSP101PTP – Item # WZ-40503-25
SPECS & DESCRIPTION
Specifications & Description
  • HeaderDANGER
  • LanguagesEnglish
  • MaterialPlastic
  • DescriptionLockout/Tagout Tag, Danger-Do Not Operate, Bilingual, 5.8x3.4\", Plastic; 25/Pk
More About this Item
Tags comply with tagout requirements of the OSHA Lockout/Tagout standard 29 CFR 1910.147. Use tags to mark and identify the lockout situation to prevent the accidental or unexpected start up or release of stored energy when working on equipment and machinery. Tags feature top gabled corners and 3/8” (9.5-mm) hole. All tags are sold in packages of 25.
Compliance: OSHA 29 CFR 1910.147
Plastic is a 15-mil rigid vinyl plastic material that is durable and will withstand tears, weathering, and various chemicals.