This item is now discontinued. Please browse similar items
Accuform MVHR633VS Safety Sign, Caution - Watch For Forklift Traffic, 10" X 14", Adhesive Vinyl
Representative image only.
Accuform Mfr # MVHR633VS – Item # WZ-51014-74
SPECS & DESCRIPTION
Specifications & Description
  • LegendWATCH FOR FORKLIFT TRAFFIC
  • HeaderCAUTION
  • LanguagesEnglish
  • MaterialAdhesive Vinyl
  • Width (in)14
  • Height (in)10
  • DescriptionSafety Sign, Caution - Watch For Forklift Traffic, 10\" X 14\", Adhesive Vinyl
More About this Item
These signs are intended to indicate potential, type, and degree of the hazard that may lead to accidental injury or property damage. Signs define specific hazards by conveying a clear, easily understood message. The sign headers and colors used in accident prevention safety signs are specified by OSHA and ANSI to designate the severity of the hazard. Each color has a specific role.
CAUTION (yellow): Indicates a potentially hazardous situation which, if not avoided, may result in minor or moderate injury.
Adhesive vinyl is a flexible 4-mil pressure-sensitive vinyl with adhesive back and peel-off liner. This material is ideal for low-cost, light-duty applications.