This item is now discontinued. Please browse similar items
Accuform MVHR633VP Safety Sign, Caution - Watch For Forklift Traffic, 10" X 14", Plastic
Representative image only.
Accuform Mfr # MVHR633VP – Item # WZ-51014-75
SPECS & DESCRIPTION
Specifications & Description
  • LegendWATCH FOR FORKLIFT TRAFFIC
  • HeaderCAUTION
  • LanguagesEnglish
  • MaterialPlastic
  • Width (in)14
  • Height (in)10
  • DescriptionSafety Sign, Caution - Watch For Forklift Traffic, 10\" X 14\", Plastic
More About this Item
These signs are intended to indicate potential, type, and degree of the hazard that may lead to accidental injury or property damage. Signs define specific hazards by conveying a clear, easily understood message. The sign headers and colors used in accident prevention safety signs are specified by OSHA and ANSI to designate the severity of the hazard. Each color has a specific role.
CAUTION (yellow): Indicates a potentially hazardous situation which, if not avoided, may result in minor or moderate injury.
Plastic is 0.055” polyethylene plastic and is good for medium duty indoor environments and sheltered outdoor environments.