This item is now discontinued. Please browse similar items
Accuform MPPE159VP Safety Sign, Danger - Respirator Required, 7" x 10", Plastic
Representative image only.
Accuform Mfr # MPPE159VP – Item # WZ-40711-94
Protect your workers by clearly displaying areas requiring a respirator
SPECS & DESCRIPTION
Specifications & Description
  • HeaderDANGER
  • LanguagesEnglish
  • MaterialPlastic
  • Width (in)10
  • Height (in)7
  • DescriptionSafety Sign, Danger - Respirator Required, 7\" x 10\", Plastic
More About this Item
These signs are intended to indicate potential, type, and degree of the hazard that may lead to accidental injury or property damage. Signs define specific hazards by conveying a clear, easily understood message. The sign headers and colors used in accident prevention safety signs are specified by OSHA and ANSI to designate the severity of the hazard. Each color has a specific role.
DANGER (red): indicates an imminently hazardous situation which, if not avoided, will result in death or serious injury.