This item is now discontinued. Please browse similar items
Accuform MFPR105VA Sign, Danger-Fall Protection Required Beyond..., 10x14", Aluminum; 1/Pk
Representative image only.
Accuform Mfr # MFPR105VA – Item # WZ-51014-07
SPECS & DESCRIPTION
Specifications & Description
  • HeaderDANGER
  • LanguagesEnglish
  • MaterialAluminum
  • Width (in)14
  • Height (in)10
  • DescriptionSign, Danger-Fall Protection Required Beyond..., 10x14\", Aluminum; 1/Pk
More About this Item
These signs are intended to indicate potential, type, and degree of the hazard that may lead to accidental injury or property damage. Signs define specific hazards by conveying a clear, easily understood message. The sign headers and colors used in accident prevention safety signs are specified by OSHA and ANSI to designate the severity of the hazard. Each color has a specific role.
DANGER (red): Indicates an imminently hazardous situation which, if not avoided, will result in death or serious injury.
Aluminum is 0.040” thick with overlaminate coat for long-term outdoor exposure. It withstands splashes and washdown without rusting.