This item is now discontinued. Please browse similar items
Accuform MEQM150VA Sign, Danger-This Equipment Starts/Stops Automatically, 7x10", Aluminum; 1/Pk
Representative image only.
Accuform Mfr # MEQM150VA – Item # WZ-50710-04
  • 5 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • HeaderDANGER
  • LanguagesEnglish
  • MaterialAluminum
  • Width (in)10
  • Height (in)7
  • DescriptionSign, Danger-This Equipment Starts/Stops Automatically, 7x10\", Aluminum; 1/Pk
  • Warranty5 years
5 Year Warranty
Key Features
  • 5 year warranty
More About this Item
These signs are intended to indicate potential, type, and degree of the hazard that may lead to accidental injury or property damage. Signs define specific hazards by conveying a clear, easily understood message. The sign headers and colors used in accident prevention safety signs are specified by OSHA and ANSI to designate the severity of the hazard. Each color has a specific role.
DANGER (red): Indicates an imminently hazardous situation which, if not avoided, will result in death or serious injury.
Aluminum is 0.040” thick with overlaminate coat for long-term outdoor exposure. It withstands splashes and washdown without rusting.