This item is now discontinued. Please browse similar items
Accuform MCLR802VA Safety Sign, Notice - Cleanroom, 10" x 14", Aluminum
Representative image only.
Accuform Mfr # MCLR802VA – Item # WZ-50711-14
Keep your cleanroom from contamination by clearly displaying locations requiring cleanroom garments
SPECS & DESCRIPTION
Specifications & Description
  • HeaderNOTICE
  • LanguagesEnglish
  • MaterialAluminum
  • Width (in)14
  • Height (in)10
  • DescriptionSafety Sign, Notice - Cleanroom, 10\" x 14\", Aluminum
More About this Item
These signs are intended to indicate potential, type, and degree of the hazard that may lead to accidental injury or property damage. They are intended to define specific hazards by conveying a clear, easily understood message. The sign headers and colors used in accident prevention safety signs are specified by OSHA and ANSI to designate the severity of the hazard. Each color has a specific role.
NOTICE (blue): Indicates a statement of company policy directly or indirectly related to the safety of personnel or protection of property.