This item is now discontinued. Please browse similar items
Accuform LZH618PS2 GHS Label, Environment Pictogram, 2" x 2", adhesive paper, 250/roll
Representative image only.
Accuform Mfr # LZH618PS2 – Item # WZ-50713-12
SPECS & DESCRIPTION
Specifications & Description
  • HeaderENVIRONMENT
  • LanguagesEnglish
  • MaterialAdhesive Paper
  • Width (in)2
  • Height (in)2
  • Quantity250/roll
  • DescriptionGHS Label, Environment Pictogram, 2\" x 2\", adhesive paper, 250/roll
More About this Item
GHS (Globally Harmonized System) is a worldwide program for classifying chemicals in the workplace. GHS recognizes nine hazard pictograms. The pictograms identify the hazard which helps users take proper precautions to minimize adverse effects.
GHS Environment pictogram indicates the following potential hazard: aquatic toxicity.