Accuform LZH608EV5 GHS Label, Environment Pictogram, 1" x 1", adhesive poly, 500/roll
Representative image only.
Accuform Mfr # LZH608EV5 – Item # WZ-50713-11
£32.75GBP / Roll of 500
£39.30GBP (incl VAT)
SPECS & DESCRIPTION
Specifications & Description
  • HeaderENVIRONMENT
  • LanguagesEnglish
  • MaterialAdhesive Poly
  • Width (in)1
  • Height (in)1
  • Quantity500/roll
  • DescriptionGHS Label, Environment Pictogram, 1" x 1", adhesive poly, 500/roll
More About this Item
GHS (Globally Harmonized System) is a worldwide program for classifying chemicals in the workplace. GHS recognizes nine hazard pictograms. The pictograms identify the hazard which helps users take proper precautions to minimize adverse effects.
GHS Environment pictogram indicates the following potential hazard: aquatic toxicity.