This item is now discontinued. Please browse similar items
Accuform KDD237 Hinge-Style Plug Lockout, for 115 V Plugs up to 3-1/4" x 1-3/4"
Representative image only.
Accuform Mfr # KDD237 – Item # WZ-40504-16
Completely closes off plug to prevent equipment from being plugged in
  • Place plug in lockout, close and secure with padlock to prevent opening of lockout device
  • Clam shell design locks the two halves together
  • Accommodates padlock with shackle diameter up to 9/32" (7.1 mm)
  • Manufactured with highly visible red plastic
  • Lockout for 115 V plugs fits sizes up to 3-1/4" x 1-3/4" (83 x 44 mm) and cords up to 1/2" (12.7 mm) diameter
SPECS & DESCRIPTION
Specifications & Description
  • DescriptionHinge-Style Plug Lockout, for 115 V Plugs up to 3-1/4\" x 1-3/4\"
Key Features
  • Place plug in lockout, close and secure with padlock to prevent opening of lockout device
  • Clam shell design locks the two halves together
  • Accommodates padlock with shackle diameter up to 9/32" (7.1 mm)
  • Manufactured with highly visible red plastic
  • Lockout for 115 V plugs fits sizes up to 3-1/4" x 1-3/4" (83 x 44 mm) and cords up to 1/2" (12.7 mm) diameter
More About this Item
Place one plug in the lockout, close, and secure with padlocks to prevent opening of the lockout. Accommodates padlocks (not included) with shackle diameters of up to 9/32" (7.1 mm). High-visibility red, all-plastic construction.
Lockout for 115 V plugs fits sizes up to 3-1/4" x 1-3/4" (8.3 x 4.4 cm) and cords up to 1/2" (1.3 cm) diameter.