This item is now discontinued. Please browse similar items
Accuform KDD122 Double-Pole Circuit Breaker Lockout, Red; Breaker Toggles Up To 5/8"
Representative image only.
Accuform Mfr # KDD122 – Item # WZ-40504-09
Lock out electrical power at the source – right at the circuit breaker, no need to lock the entire panel
  • Manufactured of rigid plastic in visible bright red
  • No modifications required, attach directly to the breaker toggle and panel to prevent the breaker from being switched on once padlock is in place
  • Fits over most major brands of breaker toggles, in the off position
  • Manufactured of rigid plastic in visible bright red
  • Accommodates padlocks with shackle diameter up to 9/32" (7.1 mm) (sold separately)
  • Double-pole style fits over breaker toggle up to 5/8" (17.7 mm) width connected by a cross bar
SPECS & DESCRIPTION
Specifications & Description
  • DescriptionDouble-Pole Circuit Breaker Lockout, Red; Breaker Toggles Up To 5/8\"
Key Features
  • Manufactured of rigid plastic in visible bright red
  • No modifications required, attach directly to the breaker toggle and panel to prevent the breaker from being switched on once padlock is in place
  • Fits over most major brands of breaker toggles, in the off position
  • Manufactured of rigid plastic in visible bright red
  • Accommodates padlocks with shackle diameter up to 9/32" (7.1 mm) (sold separately)
  • Double-pole style fits over breaker toggle up to 5/8" (17.7 mm) width connected by a cross bar
More About this Item
No need to lock out the entire panel; these compact, universally designed lockouts fit over most major brands of breaker toggles in the "off" position. Accommodates padlocks (not included) with shackle diameter up to 9/32" (7.1 mm).
Double-Pole Style fits over tow breaker toggles up to 5/8" (1.6 cm) width connected by a cross bar.