This item is now discontinued. View the direct replacement: WZ-11124-54 or browse similar items
A&D Weighing HV-200KGL Triple Resolution Bench Scale, LCD, 150/300/500 lb SKU 1112210
Representative image only.
A&D Weighing Mfr # HV-200KGL – Item # WZ-11122-10
Tilting LCD feature enhances visibility
  • Comparator for setting high and low weight targets
  • Tilting meter head for best viewing angle
  • Counting and percent function
  • 2 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • Capacity (kg)500
  • Capacity Dual Range60, 150, or 220 kg
  • Weighing Unitskg, lb, oz, %, pcs
  • Calibration TypeExternal
  • OutputRS-232
  • Battery6 x D (not included)
  • Power (VAC)115 / 230
  • DescriptionHigh Resolution Bench Scale, Lcd, 60/150/220kg (Triple Range )
  • Warranty2 Years
Two year warrantyConformite EuropeenneNTEPUL Listed.
Key Features
  • Comparator for setting high and low weight targets
  • Tilting meter head for best viewing angle
  • Counting and percent function
  • 2 year warranty
More About this Item
These scales provide counting, percentage, and comparator functions. Ideal for packaging, shipping, pharmaceutical, and industrial applications.
HV-G series of scales offer resolution that can be changed automaticlly for 3 Capacity levels.
HW-G series of scales has high resolution of 1/10,000.
V model scales have a Vacuum Fluorescent display, that is ideal for use in dark areas.
L model scales have a LCD display.