This item is now discontinued. Please browse similar items
A&D Weighing AD-1683 Static Eliminator for Use with Analytical Balances
Representative image only.
A&D Weighing Mfr # AD-1683 – Item # WZ-11110-46
Helps achieve a more stable and accurate reading by eliminating static
  • Eliminate static charge from samples by generating ions with little or no reverse charge — great for highly-electrified samples
  • Compact and lightweight with no fan and no breeze to affect the balance
  • Easy-to-use and safe to install with no high-voltage wiring
  • Use with an analytical or toploading balances
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • DescriptionStatic Eliminator for Use with Analytical Balances
  • Warranty1 year
1 Year WarrantyConformite EuropeenneUL Listed.
Key Features
  • Eliminate static charge from samples by generating ions with little or no reverse charge — great for highly-electrified samples
  • Compact and lightweight with no fan and no breeze to affect the balance
  • Easy-to-use and safe to install with no high-voltage wiring
  • Use with an analytical or toploading balances