A&D Weighing AD-1653 Density Determination Kit for BA, BM, GH, HR-I, GR, HR, and HA Series Balances
Representative image only.
A&D Weighing Mfr # AD-1653 – Item # WZ-11114-74
Use your balance to determine density with these convenient kits
  • Expand the capabilities of your balance with these convenient kits
  • Ensure the kit corresponds to your correct A&D Weighing balance
  • 3 month warranty
PRODUCT OPTIONS
£901.32GBP / Each
£1,081.58GBP (incl VAT)
Consider alternative...
SPECS & DESCRIPTION
Specifications & Description
  • DescriptionDensity Determination Kit ,
  • Warranty3 month
3 Month WarrantyConformite Europeenne
Key Features
  • Expand the capabilities of your balance with these convenient kits
  • Ensure the kit corresponds to your correct A&D Weighing balance
  • 3 month warranty
More About this Item
These density determination kits, when combined with one of the A&D Weighing electronic balances, allows the density of a solid and liquid to be measured easily. Balances sold separately.