This item is now discontinued. Please browse similar items
3M 11513-00000 1.5 Magnifier Safety Eyewear, Clear Lens
Representative image only.
3M Mfr # 11513-00000-20 – Item # WZ-86472-04
Safety glasses for those who need the assistance of a reading lens
SPECS & DESCRIPTION
Specifications & Description
  • Frame ColorBlack
  • Lens ColorClear
  • ComplianceANSI Z87.1-2003; CSA Z94.3
  • Magnification+1.5
  • Description1.5 Magnifier Safety Eyewear, Clear Lens
Canadian Standards Association
More About this Item
Large lens size offers maximum protection while allowing for reading magnification at close range. Placement of magnifying lens, like a bifocal, reduces headaches and eyestrain. Wraparound polycarbonate construction is designed to resist impacts and block 99.9% of UV rays. Flexible temples and universal-fit nosebridge.
Compliance: ANSI Z87.1-2003, CSA Z94.3