Cole-Parmer 316 Stainless Steel Turbine Propeller, 2" dia x 3/8" bore dia
Representative image only.
Cole-Parmer Item # WZ-04552-15
Keep your overhead mixer running smoothly
  • 2 year warranty
PRODUCT OPTIONS
£74.55GBP / Each
£89.46GBP (incl VAT)
SPECS & DESCRIPTION
Specifications & Description
  • Description316 Stainless Steel Turbine Propeller, 2\" dia x 3/8\" bore dia
  • Warranty2 year
Two year warranty
Key Features
  • 2 year warranty
More About this Item
Update clamps, mounts, shafts and propellers to accommodate new protocols. These replacement parts will keep your overhead mixer running smoothly.
Turbine propellers are ideal for airless mixing in deep containers.