Categories
Brand (2)
Price Range
Availability
Product Type
3A Sanitary Regulatory Approval
Application
B/T Masterflex tubing size
Barb Size-Port 2 (in)
Barb Size / Tubing Id (Port 1) (in)
All Masterflex Products
Only Masterflex peristaltic tubing pumps offer unparalleled and standard-setting reliability—performance produced by more than 60 years of manufacturing peristaltic pumps. Masterflex tubing pumps offer flow rates as low as 0.00001 mL/min to 42 LPM and are able to generate pressures up to 10 bar (150 psi). They are easy to use and, in most cases, can be installed within minutes. With a few moving parts, they are also easy to maintain. With many tubing formulations available, you can ensure compatibility with an extensive variety of fluids. From low-cost fixed-speed pumps to advanced remote-controlled models suitable for critical metering and dispensing applications, we can provide you with a tubing pump solution.
APPLY FILTERS | 2
filter byclear all
806 RESULTS
SORT BY
Showing 1 - 24 of 806
2334>