Ease sample preparation and analysis of surface lead or surface dust with GhostWipes. Made of sturdy material moistened with DI water, the wipes hold together even on the roughest surfaces.
In the lab, the GhostWipe completely dissolves during the digestion process. This provides complete dispersion of analytes and uniform recoveries—no messy fibrous material to clog the sample uptake capillary or nebulizer.
Watch the video below!
Thanks for a great information .we got lot of information from your article and soon we waiting for the next update.. Go-Green