We name seven of our Masterflex L/S drives for pumping efficiency. Masterflex® L/S® Drives—one of our most popular pumps—is available in various speed types and formats. Which works best for your application?
1) Fixed-Speed (flow rates: 0.06 to 290 mL/min)
- Consistent repeatable flow rates
- Economical and compact
- UL, cUL, and CE listed models
2) Variable-Speed Economy Console (flow rates: 0.42 to 2900 mL/min)
- Economical variable flow
- Simple speed control
- UL, cUL, and CE listed models
- Accept stacked pump heads for multiple channels
3) Precision Modular (flow rates: 0.06 to 3400 mL/min)
- Separate motor and controller up to 15 ft (4.6 m)
- Models available with IP66-rated wall-mount controllers
- Pump heads can be stacked
4) Precision Variable-Speed Console (flow rates: 0.06 to 3400 mL/min)
- Economical variable flow
- Simple speed control
- ETL and CE listed
- Accept stacked pump heads for multiple channels
- Drives are stackable
5) Variable-Speed Digital and Computer Compatible (flow rates: 0.001 to 3400 mL/min)
- Four-line graphical LCD with menu-driven programming
- Maintenance-free brushless motor
- PWM speed control with 6000:1 turndown
- Tachometer feedback for ±0.1% speed control
- Calibration capability
6) Digital Process (flow rates: 0.006 to 3400 mL/min)
- Four-line graphical LCD with menu-driven programming
- Sealed housing; IP66 and NEMA 4X rated for washdown
- Maintenance-free brushless motor; PWM speed control
- Dispensing features include delay interval and cumulative volume
- Remote control capability
7) Digital Modular Dispensers (flow rates: 0.001 to 3400 mL/min)
- Maintenance-free brushless motor
- Precise repetitive dispensing with antidrip
- Precise PWM speed control
- Quiet operation
- Tachometer feedback for ±0.1% speed control
- Available with washdown wall-mount controller
- Calibration capability
Be the first to comment on "7 Masterflex L/S Drives for Pumping Efficiency"