Onguard
Onguard
On Guard acid detecting paint protects employees, equipment, and the environment. The paint changes color to alert you to the presence of acids. Choose from one gallon paint or trigger spray bottles. We are an On Guard authorized distributor.