Bacti-Cinerator
Bacti-Cinerator
Bacti-Cinerator IV Sterilizer takes only 5 to 7 seconds to sterilize needles and platinum loops. Infrared heat is deep within the ceramic funnel to protect lab staff from an open flame. We are a Bacti-Cinerator authorized dealer.