Cole-Parmer - The world's leading source for scientific and industrial instrumentation since 1955.
United Kingdom   Cart  Subtotal = 0.00
0500 345 300 Items = 0  View Cart
Login Login
New Customer? Register
Home Products & Services Technical Library myCP Account Contact/About Us
Cole-Parmer UK Catalog > Pumps > Laboratory Pumps > Vacuum Pressure Pumps 
Vacuum Pressure Pumps
  More Information Buy
Drum Drum pumps Shop online
Peristaltic Peristaltic pumps Shop online
Gear Gear pumps Shop online
Diaphragm Diaphragm pumps Shop online
Piston Piston pumps Shop online
Syringe Syringe pumps Shop online
Centrifugal Centrifugal pumps Shop online
Air Cadet Vacuum/Pressure Pump
• Low cost diaphragm vacuum pump
• Max vacuum: 780 mbar (584 Torr)
Vacuum Pumps
• Free-air capacity: 13 LPM
from  Call for price   More details
Standard Duty Dry Vacuum Piston Pumps
• Great for filtering, aspirating, degassing and desiccating
• Max vacuum: 80 mbar (60 Torr)
dry vacuum pump
• Free-air capacity: 38 LPM
from  Call for price   More details
VacuuBrand ® Chemical-Resistant Diaphragm
• Chemically resistant diaphragm pump
• Max vacuum: 4 mbar (3 Torr)
chemical resistant pump
• Free-air capacity: 34 LPM
from  Call for price   More details
Cole-Parmer offers a complete line of vacuum pumps for laboratory, environmental sampling, process and OEM use. From free-standing vacuum pumps to self-regulating pumping systems Cole Parmer UK offers vacuum pump technologies to cover a wide range of ultimate vacuum down to 1 x 10-8 mbar (7.5 x 10-9 Torr).
Use the vacuum pump selection guide below and browse through our on-line catalogue to find your vacuum pressure pump, alternatively contact our Application Specialists for free technical support.
Vacuum Pump Selection Guide
The technology chosen for vacuum pressure pump applications is largely dependent on the desired vacuum level:
  • For ultimate vacuums down to 0.3 mbar (0.2 Torr) consider:
    • Diaphragm Vacuum Pumps
    • Piston Vacuum Pumps
    • Venturi Vacuum Pumps
  • For ultimate vacuums down to 2 x 10-3 mbar (1.5 x 10-3 Torr) consider:
    • Rotary Vane Vacuum Pumps
  • For ultimate vacuums down to 1 x 10-8 mbar (7.5 x 10-9 Torr) consider:
    • Turbomolecular Vacuum Pumps
When selecting a vacuum pump in addition to the ultimate vacuum required consideration also needs to be given to:
  • Chemical compatibility - Assess the compatibility of the gases with the pump and any potential problems with the process gas (eg moisture). Cole-Parmer offers many chemically resistant options for highly corrosive applications.
  • Pumping speed (volume flow) and pressure requirements – Assess the vacuum pump’s capability to produce both the required pumping speed (volume flow) and throughput (mass flow) against the process’s needs by using the equation Q (gas load) = S (pumping speed) x P (pressure) calculation.
  • Pump Installation – Including port connections and requirement for ancillary equipment such as a roughing pump.
  • Pump Maintenance – Including frequency of maintenance cycles and required level of expertise to carry out the service.
  • Costs – Consideration should not only be given to the initial purchase price of the vacuum pump but also the operating costs, maintenance cycles and cost.
Privacy Statement  |  Terms and Conditions  |  Careers@Cole-Parmer  |  Press Releases  |  Site Map
Cole-Parmer Sites
Cole-Parmer: United States
  Canada
  China
  United Kingdom
  India
MasterFlex.com - Ordering and Technical Info
FoodTechSource.com - Food Quality Site
InnocalSolutions.com - Calibration and Certification
Cole-Parmer
Unit 3 River Brent Business Park
Trumpers Way, Hanwell, London W7 2QA
Telephone: 020 8574 7556
Fax: 020 8574 7543
E-Mail: sales@Coleparmer.co.uk
Copyright © 2007