Cole-Parmer - The world's leading source for scientific and industrial instrumentation since 1955.
United Kingdom   Cart  Subtotal = 0.00
0500 345 300 Items = 0  View Cart
Login Login
New Customer? Register
Home Products & Services Technical Library myCP Account Contact/About Us
Cole-Parmer UK Catalog  > Tubing 
Transfer Tubing
  More Information Buy
flexible Flexible Tubing Shop online
heat Heat Shrink Tubing Shop online
Microbore Microbore Tubing Shop online
Rigid Rigid Tubing Shop online
Tubing Tubing Accessories Shop online
Silicone (peroxide-cured) tubing - 1/8'' ID x 1/4''OD - 25 ft/pack
• Sterilize: With autoclave or gamma irradiation
• Temperature range: –60 to 460°F (–51 to 238°C)
Silicone tubing
• Characteristics: Excellent heat resistance, low compression set, odorless, nontoxic. Translucent
from  Call for price   More details
PharMed BPT tubing - 1/4'' ID x 1/2'' OD - 500ft/pack
• Ideal for applications including cell and tissue culture work
• Temperature range: –75 to 275°F (–59 to 135°C)
Pharmed Tubing
• Characteristics: nontoxic, opaque beige, extremely low permeability
from  Call for price   More details
PTFE tubing - 1/32'' ID x 1/16'' OD - 25 ft/pack
• Sterilize: With autoclave, ethylene oxide, or use dry heat
• Temperature range: -400 to 500°F (-240 to 260°C)
ptfe tubing
• Characteristics: Chemically inert, nontoxic, and virtually nonporous. Excellent crack- and stress-resistance. Low coefficient of friction. Translucent
from  Call for price   More details
Cole Parmer UK offer an extensive range of transfer tubing. Our flexible and rigid tubing formulations cover a wide range of characteristics to fulfil all your transfer tubing needs. Whether it is for everyday laboratory tubing applications or for industrial transfer applications we offer:
  • Tubing Size with inside diameters as small as 0.002” (0.05 mm) to 2”(50.8mm)
  • Pressure ratings up to 30 bar
  • Tubing to withstand Vacuum level of 759mm (29.9”) of mercury at 23°C
  • Temperature ratings up to 288°C
  • Chemical compatible transfer tubing for the food, drink and pharmaceutical industry with our range of FDA approved material as well as tubing for aggressive chemicals and solvents.
  • Tubing suitable for sterilization techniques such as gamma irradiation, autoclaving and ethylene oxide sterilization
  • Tubing Resin Certification which available free of charge at time of order for a wide selection of our tubing. The certification states the tubing orders complies with regulatory standards.
  • Quantities for general use as well as spools of up to 150 m (500ft) for some our more popular ranges for production lines.
When selecting transfer tubing for your application, use our online technical tools or contact our application specialist to help you find the right tubing. We have over 50 years of experience and are always testing new liquids and gases for improved chemical compatibility ratings.
Cole-Parmer’s Popular Transfer Tubing:
Silicone tubing
Peroxide-Cured Silicone Tubing
  • Flexible, soft and translucent tubing formulations
  • Available in platinum, peroxides cured and reinforced tubing formulations
  • Able to withstand temperatures up to 232 °C
  • Sizing ranging from as small as 0.64 mm ID to 38.2 mm ID
  • Excellent biocompatibility with no leachable additives
  • FDA compliant making it ideal for pharmaceutical, biotech and food application
  • Available is standard pack sizes or spooled rolls for production runs
Tygon Tubing
Tygon lab tubing
  • Available in a range of formulations including Tygon Chemical, Tygon UV-Resistant, Tygon Vacuum and Tygon Fuel & Lubricant
  • Flexible tubing available in sizing from 0.19 mm to 38.1 mm
  • Temperature rating of up to 74 °C
  • Long tubing life formulations available
  • Applications include general laboratory use, gasoline, hydrocarbons and alcohols
  • Available is standard pack sizes or spooled rolls for production runs
Viton Tubing
Masterflex Viton tubing
  • Ideal for high temperature work up to 204 °C
  • Available in opaque white or opaque black
  • FDA approved tubing
  • Sizes from 0.89 mm ID to 19.1 mm
  • Ideal for transferring solvents and acids
  • Good chemical resistance
PTFE tubing
PTFE Tubing
  • Chemically inert, non-toxic and virtually nonporous
  • Temperature range of -240 °C to 260 °C
  • Ultra microbore tubing available 0.05 mm to 19 mm
  • FDA compliant tubing
  • Ideal for demanding lab, chemical, electrical and automotive applications
  • Translucent
  • Available is standard pack sizes or spooled rolls for production runs
Privacy Statement  |  Terms and Conditions  |  Careers@Cole-Parmer  |  Press Releases  |  Site Map
Cole-Parmer Sites
Cole-Parmer: United States
  Canada
  China
  United Kingdom
  India
MasterFlex.com - Ordering and Technical Info
FoodTechSource.com - Food Quality Site
InnocalSolutions.com - Calibration and Certification
Cole-Parmer
Unit 3 River Brent Business Park
Trumpers Way, Hanwell, London W7 2QA
Telephone: 020 8574 7556
Fax: 020 8574 7543
E-Mail: sales@Coleparmer.co.uk
Copyright © 2007