Cole-Parmer - The world's leading source for scientific and industrial instrumentation since 1955.
United Kingdom   Cart  Subtotal = 0.00
0500 345 300 Items = 0  View Cart
Login Login
New Customer? Register
Home Products & Services Technical Library myCP Account Contact/About Us
Cole-Parmer UK Catalog  > Tubing > Silicone Tubing 
Silicone Tube & Tubing
  More Information Buy
colour Silicone Color-Coded Tubing Shop online
silicone Silicone Tubing Shop online
peroxide Peroxide-Cured Reinforced Silicone Tubing Shop online
Platinum-Cured Platinum-Cured Silicone Tubing Shop online
Peroxide-Cured Silicone Tubing
• 3/8''ID x 1/2''OD - 25 ft/pack
• Applications: Pharmaceutical, biotechnology, and aerospace applications.
Peroxide cured silicone
• Characteristics: Excellent heat resistance, low compression set, odorless, nontoxic. Translucent.
from  £35.00   More details
Platinum-Cured Silicone Tubing
• 1/50''ID x 1/12''D - 25 ft/pack
• Applications: Pharmaceutical, biotechnology, and aerospace applications.
platinum cured silicone
• Characteristics: Excellent heat resistance, low compression set, odorless, nontoxic. Translucent.
from  £19.99   More details
Reinforced silicone (platinum-cured) tubing
• Applications: Demanding sanitary high-pressure pharmaceutical, biotechnology, food and beverage, and cosmetic applications
• Characteristics: Excellent heat resistance, low compression set, odorless, and nontoxic. Translucent
Reinforced silicone tubing
• Temperature range: –112 to 320°F (–51 to 204°C)
from  £731.00   More details
Platinum-Cured Silicone Tubing Spools
• Spools are sold in 300 ft or 500 ft spools.
• Reduce unpacking time and save money on shipping
Spooled tubing
• Available in Norprene ® , PharMed ® , platinum-cured silicone, or Tygon ® Lab formulations
from  £662.00   More details
Cole Parmer UK has over 50 years of experience in silicone tubing solutions. From 0.64mm (0.025”) to 38mm (1 ˝”) inside diameter (ID) we offer a number of silicone formulations and colour coding options to suit you application. Tubing is supplied in convenient 7.6m pack sizes for general use or larger spool sizes of between 90 and 150m for production size runs. All silicone tubing is lot traceable, made to food and drug administration (FDA) standards and we offer free certificates of conformity and free tubing resin compliance certificates if requested at time of order.
Select from our general purpose peroxide silicone tubing, high purity platinum cured silicone tubing or for high pressure applications our reinforced silicone tubing. For peristaltic pump applications use Masterflex peroxide cured silicone tubing or Masterflex platinum cured silicon tubing which is extruded to very tight tolerances to ensure repeatable and high accuracy delivery. When using multi channel configurations consider using our colour coded silicone tubing to provide easy identification.
Whether a liquid or gas application, high accuracy dispensing or high volume flow, Cole-Parmer can deliver tubing formulations to meet your application needs. To find the right tubing for your application use Cole-Parmer’s on-line nominal tubing quick search and chemical compatibility table to ensure correct compatibility with your media. Alternatively contact our Application Specialists for free technical support to help solve your application needs.
Key Points for silicone tubing
  • Wide range of sizing and formulations available in silicone tubing (see tables below for examples)
  • Free certificates of tubing resin compliance available upon request
  • To ensure proper selection of silicone tubing, one-foot sample lengths are available for most formulations
  • Quantity discounts available for many silicone tube formulations
  • Often custom lengths can be offered, contact our application team for any special order requirements
  • Available in pack lengths of 7.6m or spool lengths between 90m and 150m as well as bulk packs of 10 x 15 m
Advantages for silicone tubing
  • Excellent biocompatibility
  • No taste imparted to transported fluids
  • Extremely good over a wide range of temperatures
  • Weather, ozone, corona and radiation resistant
  • FDA 21 CFR 177.2600 classified
Disadvantages for silicone tubing
  • Not suitable for use with concentrated acids and bases, organic solvents or oils
  • Relatively high gas permeability
Typical industries for silicone tubing
  • Pharmaceutical
  • Biotechnology
  • Food and Beverage
  • Aerospace
  • Cosmetic
  • General laboratory use
Silicone Tubing Formulations available at Cole-Parmer
Silicone tube formulation
Temperature range (oC)
Max Bar ratings at 21oC
Characteristics
Peroxide Silicone Tubing
-51 to 238
Up to 2.1
  • Peroxide cured
  • Excellent heat resistance
  • Low compression set
  • Odorless
  • Nontoxic
  • Translucent
Platinum Silicone Tubing
-50 to 230
Up to 2.1
  • Platinum cured
  • Excellent heat resistance
  • Low compression set
  • Odorless
  • Nontoxic
  • Translucent
Tygon BioPharm Silicone Tubing
-80 to 200
Up to 1.1
  • Ultra-smooth inner surface reduces particle entrapment and minimises bacterial growth
  • Good for mild to medium concentration bases, salts & alcohols
  • Odorless, tasteless & nontoxic
  • Transparent
Platinum-Cured Reinforced Silicone Tubing
-51 to 160
Up to 21.5
  • Excellent heat resistance
  • Low compression set
  • Odorless
  • Nontoxic
  • Translucent
  • High-pressure
Peroxide-Cured Reinforced Silicone Tubing
-51 to 204
Up to 12
  • Excellent heat resistance
  • Low compression set
  • Odorless
  • Nontoxic
  • Low tackiness
  • Translucent
Colour-Coded Silicone Tubing
-50 to 230
Up to 1.4
  • High temperature tolerance
  • Opaque
  • Odorless
  • Nontoxic
Masterflex Silicone Tubing
-73 to 232
Up to 2.1 bar
  • For use with Masterflex peristaltic pumps
  • Available in C/L, L/S, I/P and B/T range
Click Here for Examples of Silicone Tubing Sizing available at Cole-Parmer
Privacy Statement  |  Terms and Conditions  |  Careers@Cole-Parmer  |  Press Releases  |  Site Map
Cole-Parmer Sites
Cole-Parmer: United States
  Canada
  China
  United Kingdom
  India
MasterFlex.com - Ordering and Technical Info
FoodTechSource.com - Food Quality Site
InnocalSolutions.com - Calibration and Certification
Cole-Parmer
Unit 3 River Brent Business Park
Trumpers Way, Hanwell, London W7 2QA
Telephone: 020 8574 7556
Fax: 020 8574 7543
E-Mail: sales@Coleparmer.co.uk
Copyright © 2007