Table scrolls horizontally
PVC and CPVC Piping Selection Guide Solvent-Welded Pressure Rating vs Service Temperature for PVC and CPVC Pipes
Nom. size |
D Outside dia. |
† Wall |
DR=D/† | Pressure rating (P) | | 73°F | 90°F | 100°F | 110°F | 120°F | 130°F | 140°F | 160°F | 180°F | 200°F | 210°F | PVC f=1 S=2000 | CPVC f=1 S=2000 | PVC f=0.75 S=1500 | PVC f=0.62 S=1240 | PVC f=0.50 S=1000 | PVC f=0.40 S=800 | CPVC f=0.65 S=1300 | PVC f=0.30 S=600 | CPVC f=0.57 S=1135 | PVC f=0.22 S=440 | CPVC f=0.50 S=1000 | CPVC f=0.40 S=800 | CPVC f=0.25 S=500 | CPVC f=0.20 S=400 | CPVC f=0.16 S=320 | | 1/2 | 0.840 | 0.147 | 5.714 | 848 | 848 | 636 | 526 | 424 | 339 | 552 | 255 | 484 | 187 | 424 | 339 | 212 | 170 | 136 | | 3/4 | 1.050 | 0.154 | 6.818 | 688 | 688 | 516 | 426 | 344 | 275 | 447 | 206 | 392 | 151 | 344 | 275 | 172 | 138 | 110 | | 1 | 1.315 | 0.179 | 7.346 | 630 | 630 | 473 | 390 | 315 | 252 | 410 | 189 | 359 | 139 | 315 | 252 | 158 | 126 | 101 | | 1-1/4 | 1.660 | 0.191 | 8.691 | 520 | 520 | 390 | 322 | 260 | 208 | 338 | 156 | 296 | 114 | 260 | 208 | 130 | 104 | 83 | | 1-1/2 | 1.900 | 0.200 | 9.500 | 471 | 471 | 353 | 292 | 235 | 188 | 306 | 141 | 268 | 104 | 235 | 188 | 118 | 94 | 75 | | 2 | 2.375 | 0.218 | 10.894 | 404 | 404 | 303 | 251 | 202 | 162 | 263 | 121 | 230 | 89 | 202 | 162 | 101 | 81 | 65 | | 2-1/2 | 2.875 | 0.276 | 10.417 | 425 | 425 | 319 | 263 | 212 | 170 | 276 | 127 | 242 | 93 | 212 | 170 | 106 | 85 | 68 | | 3 | 3.500 | 0.300 | 11.667 | 375 | 375 | 281 | 233 | 188 | 150 | 244 | 113 | 214 | 83 | 188 | 150 | 94 | 75 | 60 | | 4 | 4.500 | 0.337 | 13.353 | 324 | 324 | 243 | 201 | 162 | 130 | 210 | 97 | 185 | 71 | 162 | 130 | 81 | 65 | 52 | | 6 | 6.625 | 0.432 | 15.336 | 279 | 279 | 209 | 173 | 140 | 112 | 181 | 84 | 159 | 61 | 140 | 112 | 70 | 56 | 45 | | 8 | 8.625 | 0.500 | 17.250 | 246 | 246 | 185 | 153 | 123 | 98 | 160 | 74 | 140 | 54 | 123 | 98 | 62 | 49 | 39 | | P | = | 2S† D - † | = | 2S DR - 1 | = | P73°Ff
| = Pressure rating of pipe at service temperatures (psi) S = Hydrostatic design stress (psi) D = Outside diameter of pipe (inches) † = Pipe wall thickness (inches) f = Derating factor for service temperature DR = Dimensions ratio (D/†) P73°F = Pressure rating at 73°F Note 1: Figures for pressure rating at 73°F are rounded off from actual calculated values. Pressure ratings for other temperatures are calculated from 73°F values. Note 2: Pressure ratings are for PVC (12454-B) and CPVC (23447-B) pipes. For most sizes, pressure ratings are calculated from the experimentally determined long-term strength of PVC type 1 and CPVC extrusion compounds. Because molding compounds may differ in long-term strength and elevated temperature properties from pipe compounds, piping systems consisting of extruded pipe and molded fittings may have lower pressure ratings than those shown here, particularly at the higher temperatures. Caution should be exercised when designing PVC systems operating above 100°F and CPVC systems operating above 180°F. Note 3: The pressure ratings given are for solvent-cemented systems. When adding valves, flanges, or other components, the system must be derated to the rating of the lowest component. (Pressure ratings: molded or cut threads are rated at 50% of solvent-cemented systems; flanges and unions are 150 psi; for valves, see manufacturer's recommendation.) |