Cole-Parmer - The world's leading source for scientific and industrial instrumentation since 1955.
United Kingdom   Cart  Subtotal = 0.00
0500 345 300 Items = 0  View Cart
Login Login
New Customer? Register
Home Products & Services Technical Library myCP Account Contact/About Us
Cole-Parmer UK Catalog > Pumps > Metering pumps > Tubing Pumps 
Tubing Pumps
  More Information Buy
Drum Drum pumps Shop online
Peristaltic Peristaltic pumps Shop online
Gear Gear pumps Shop online
Diaphragm Diaphragm pumps Shop online
Piston Piston pumps Shop online
Syringe Syringe pumps Shop online
Centrifugal Centrifugal pumps Shop online
Masterflex Peristaltic Pumps
• Complete dispensing systems with Tygon LFL tubing – up and running within minutes!
• Flow rate 28 to 1,700 ml/min
Peristaltic pump system
• Includes remote control options
from  Call for price   More details
Ismatec High-Accuracy Low Flow Pumps
• Up to 24 high accuracy channels in one pump
• Analogue input and output controls
ismatec pump
• Flow rate per channel - 0.0012 to 8.86ml/min
from  £1790.00   More details
Masterflex C/L Varible - Speed Tubing Pumps
• Flow range: 0.001 to 39.4 ml/min depending on rpm and tubing size
• Remove control connections on back of pump; start/stop pump with contact closure
CL pumps
• Built-in tubing retainers firmly grip the tubing - change tubing quickly and easily
from  £305.00   More details
Masterflex I/P Digital Brushless Process Drive
• Dispense by volume or time or pump continuously
• Flow range: 0.01 to 26 LPM depending on rpm and tubing size
IP drive
• Brushless motor virtually eliminates maintenance
from  £2450.00   More details
Cole-Parmer UK offers a range of tubing pumps suitable for dispensing, metering and general transfer applications. Our tubing pumps offer flow rates as low as 0.0007 mL/min to 45 LPM and are able to generate pressures up to 8.6 bar (125 psi).
Tubing pumps confine the media to the tubing providing you with a pump that cannot contaminate the fluid and the fluid cannot contaminate your pump. This contamination free pumping makes tubing pumps great for use in high purity application including the transfer or dosing of chemicals and additives in food, pharmaceutical and semiconductor applications. In addition as the media is confined to the tubing a tubing pump offers easy maintenance and reduced down time compared to other pumping technologies.
Tubing pumps use flexible tubing to run through rollers in the pump head. As fluid goes through the pump head, packets of fluids are formed. Flow rate is determined by the size of the packets and the speed of the rollers turning. By controlling the size of the tubing and the speed of the pump head accurate metering of the media can be achieved.
Cole-Parmer offer tubing pumps suitable for use in labs, pilot plants, process and OEM applications. From low cost fixed speed pumps to advanced controlled models suitable for critical metering and dispensing applications Cole-Parmer is able to offer a tubing pump solution.
Browse through our on-line catalogue and use our tools to find your tubing pump or contact our application specialist for free technical support and let us bring over 50 years of pumping experience to solve your application needs.
Advantages for tubing pumps
  • The only part of a tubing pump that comes in contact with the liquid is the tubing. This maintains the sterility of fluid and the pump
  • Cole-Parmer offers over 20 tubing formulations and sizes for their tubing pumps for excellent compatibility with a variety of liquids
  • Self priming – our tubing pump will draw fluid into the tubing when starting dry, up to 8.8 m suction lift. Other pumps require user to fill pump and suction line with fluid before use which can be inconvenient and also create potential for hazardous spills or contamination
  • Gentle pumping action – low shear – makes a tubing pump ideal for handling cells and large proteins, as the pumping action will not damage the content
  • The fluid does not contact the tubing pump so there are no parts to clean such as valves, diaphragms, seals etc. All you have to do is clean or replace the tubing. This easy tubing replacement means reduced maintenance times
  • A tubing pump is non-siphoning which means that they prevent back flow into the system. This promotes accuracy during dispensing
Typical Application for tubing pumps
  • Juice production
  • Pizza sauce dispensing
  • Vitamin A & D Injection
  • Aspiration of tissue culture medium
  • Circulation of cell suspension in fermentation
  • Cosmetic dispensing
  • Harvesting cell media
  • Manufacturing IV bag dispensing
  • Nutrient supply for cultures
  • Spray coating dispensing
  • Sterilized media dispensing
  • Ultrafiltration
  • Acid/base dispensing
  • Adhesives for cement
  • Circuit board manufacturing
  • Dispensing glue emulsions
  • Transfer of Fuels and lubricants
Privacy Statement  |  Terms and Conditions  |  Careers@Cole-Parmer  |  Press Releases  |  Site Map
Cole-Parmer Sites
Cole-Parmer: United States
  Canada
  China
  United Kingdom
  India
MasterFlex.com - Ordering and Technical Info
FoodTechSource.com - Food Quality Site
InnocalSolutions.com - Calibration and Certification
Cole-Parmer
Unit 3 River Brent Business Park
Trumpers Way, Hanwell, London W7 2QA
Telephone: 020 8574 7556
Fax: 020 8574 7543
E-Mail: sales@Coleparmer.co.uk
Copyright © 2007