Cole-Parmer - The world's leading source for scientific and industrial instrumentation since 1955.
United Kingdom   Cart  Subtotal = 0.00
0500 345 300 Items = 0  View Cart
Login Login
New Customer? Register
Home Products & Services Technical Library myCP Account Contact/About Us
Cole-Parmer UK Catalog > Pumps > Metering Pumps 
Metering Pumps
  More Information Buy
Drum Drum pumps Shop online
Peristaltic Peristaltic pumps Shop online
Gear Gear pumps Shop online
Diaphragm Diaphragm pumps Shop online
Piston Piston pumps Shop online
Syringe Syringe pumps Shop online
Centrifugal Centrifugal pumps Shop online
Masterflex Peristaltic Pumps
• Complete dispensing systems with Tygon LFL tubing – up and running within minutes!
• Flow rate 28 to 1,700 ml/min
Peristaltic pump system
• Includes remote control options
from  Call for price   More details
Diaphragm Metering Pump
• Rugged, chemically resistant casing with choice of wetted parts
• Select from manual, remote or digital control models
Diaphragm metering pump
• Flow ranges: Liquid 0.4 ml/min – 76 LPH
from  Call for price   More details
Digital Gear Pump System
• Flow rate up to 4212 ml/min available
• Calibrate, copy and dispense features
digital gear pump
• Remote control capabilities by voltage and current
from  Call for price   More details
Cole-Parmer Syringe Pumps
• Ideal for low-flow, low pressure precision applications
• Flow rates up to 500 ml/hr
syringe pump
• Programmable models available
from  £990.00   More details
Piston Pump Heads
• High accuracy - better than +/- 1%
• Flow rates up to 1035 ml/min available
piston pump
• Order drives seperately
from  £193.00   More details
Metering pumps offer a high degree of accuracy and are used in a wide variety of applications where precise volumes of liquid are required to be moved in a specified time. A metering pump is a subset of the positive displacement pump type and discharges a known volume with every revolution or cycle. Cole-Parmer’s range of metering pumps can cover flow rates from 1 nL/min to 225 LPM with pressure options up to 345 bar.
Cole-Parmer offers a range of metering pump categories, including diaphragm pumps, gear pumps, peristaltic pumps and piston pumps. With options for laboratory, pilot plant, industrial and OEM applications browse through our on-line catalogue. Alternatively contact our Application Specialists for free technical support and let us bring over 50 years of pumping experience to help solve your application needs.
A metering pump transfers media in two phases:
  • The intake stroke where the inlet side of the pump is opened, the outlet side closed and liquid is pulled in to the pump cavity
  • The outlet stroke where the inlet side of the pump is closed, the outlet side opened and liquid is forced out
Metering pumps are generally used in applications where one or more of the following conditions apply:
  • High accuracy dosing or movement of media is required
  • Pumping need to be controlled by computer, PLC, or flow proportioning
  • Flow rates are LPH or less
  • High system pressure
  • High viscosity or slurry media
Image1
Diaphragm Pumps
Widely used in industrial applications including dosing of chemicals in process applications, water and waste water treatment
  • Flow: 0.4 ml/min to 8.3 LPM
  • Max Pressure: 17.3 bar
  • Wide range of control options available including manual, analogue, flow proportioning, computer or PLC
  • ATEX versions available
  • Robust pumps capable of delivering high accuracy and working for long periods between maintenance
Image2
Gear Pumps
Used in industrial, OEM and laboratory applications including chemical processing, paints and ink industries
  • Flow: 0.1 ml/min to 225 LPM
  • Max Pressure: 6.8 bar
  • Wide range of control options available including manual, analogue, flow proportioning, computer or PLC
  • ATEX options available
  • Offers accurate, pulseless flow and with the Micropump range a magnetically coupled pump head to deliver leak-free pumping
Image3
Peristaltic Pumps
Widely used in laboratory, process and OEM applications including chemical, food, pharmaceutical, semiconductor and water treatment industries
  • Flow: 0.0006 ml/min to 45 LPM
  • Max Pressure: 8.6 bar
  • Wide range of control options available including manual, analogue, computer or PLC control via RS-232
  • ATEX versions available
  • Media is confined to the tubing allowing contamination free pumping and easy maintenance, minimising down time.
Image4
Piston Pump
Widely used in laboratory, process and OEM applications for high accuracy delivery of media including HPLC pumping.
  • Flow: 0.04 ml/min to 1.3 LPM
  • Max Pressure: 345 bar
  • Wide range of control options available including manual, analogue, computer or PLC control via RS-232
  • Highly accurate over long periods of operation and provide excellent repeatability
Image5
Syringe Pump
Widely used in laboratory for nanofluidic and microfluidic infusion and withdrawal applications
  • Flow: 1 nL/min to 147 ml/min
  • Wide range of control options available including manual, analogue, computer or PLC control via RS-232.
  • Provides high accuracy delivery of ultra low flows
Privacy Statement  |  Terms and Conditions  |  Careers@Cole-Parmer  |  Press Releases  |  Site Map
Cole-Parmer Sites
Cole-Parmer: United States
  Canada
  China
  United Kingdom
  India
MasterFlex.com - Ordering and Technical Info
FoodTechSource.com - Food Quality Site
InnocalSolutions.com - Calibration and Certification
Cole-Parmer
Unit 3 River Brent Business Park
Trumpers Way, Hanwell, London W7 2QA
Telephone: 020 8574 7556
Fax: 020 8574 7543
E-Mail: sales@Coleparmer.co.uk
Copyright © 2007