Cole-Parmer - The world's leading source for scientific and industrial instrumentation since 1955.
United Kingdom   Cart  Subtotal = 0.00
0500 345 300 Items = 0  View Cart
Login Login
New Customer? Register
Home Products & Services Technical Library myCP Account Contact/About Us
Cole-Parmer UK Catalog > Pumps > Hand and Drum Pumps 
Hand Pumps and Drum Pumps
  More Information Buy
Peristaltic Peristaltic pumps Shop online
Gear Gear pumps Shop online
Diaphragm Diaphragm pumps Shop online
Piston Piston pumps Shop online
Syringe Syringe pumps Shop online
Centrifugal Centrifugal Pumps Shop online
Siphon/Lift Pail Pump
• Suitable for pumping petroleum products or water
• Cut inlet tube to fit your container
hand operated pump
• Maximum suction lift is 10ft
from  Call for price   More details
Electric Drum Motor
• Build a pump best suitable for your application - starting with this 9000 rpm motor
• Use with our tube sets that can handle viscosities to 50,000 cp
Drum motor
• Corrosion resistant expoxy coating resists caustic fluids
from  Call for price   More details
Mixing Drum Pumps
• Air operated drum pumps
• Mix and pump emulsions and suspensions in a single operation
pump
• Small pump tube diameter is ideal for use in applications with narrow necked containers
from  Call for price   More details
Cole-Parmer UK offers an extensive range in economical hand-operated pumps as well as electric driven and air-operated drum pumps. They are ideal for transferring fluids from drums or other containers up to 200 litres in capacity. Our range of hand and drum pumps are able to deliver flow rates of up to 238 LPM and pressures of 6.8 bar. They are ideal for delivering pulseless flow and are able to pump more viscous fluids up to 100,000 cp.
Hand pumps – Cole-Parmer offers several designs in hand operated pumps including siphon, piston, diaphragm and rotary hand-operated drum pumps. They are available in a range of materials from low cost plastics to stainless steel and PTFE, making them ideal for transferring non-corrosive chemicals such as oils, waxes and soaps as well as corrosives, detergents and acids.
Drum pumps – Cole-Parmer offers air operated as well as electric driven drum pumps. Our drum pump systems have a range of varying characteristics from short to long tube lengths for small to large containers, tube materials including plastic, PVDF, sanitary and stainless steel, motors that are explosion proof or air operated and batch controlled drum pumps to allow automatic dispensing of preset volumes.
Use Cole-Parmer’s on-line catalogue to find and compare pump specifications. Alternatively contact our Application Specialists for free technical support and let us bring over 50 years of pumping experience to help solve your application needs.
Advantages for hand and drum pumps
  • Different tube set materials to best suit your needs and applications
  • Adapters available for a range of containers
  • Options include variable speed motors, air operated and explosion proof motors
Typical Application for hand and drum pumps
  • Acid & Alkali drum pumps
  • Sanitary maintenance drum pumps
  • Petroleum drum pumps
  • Water & Utility pumps
Privacy Statement  |  Terms and Conditions  |  Careers@Cole-Parmer  |  Press Releases  |  Site Map
Cole-Parmer Sites
Cole-Parmer: United States
  Canada
  China
  United Kingdom
  India
MasterFlex.com - Ordering and Technical Info
FoodTechSource.com - Food Quality Site
InnocalSolutions.com - Calibration and Certification
Cole-Parmer
Unit 3 River Brent Business Park
Trumpers Way, Hanwell, London W7 2QA
Telephone: 020 8574 7556
Fax: 020 8574 7543
E-Mail: sales@Coleparmer.co.uk
Copyright © 2007